Learn More
Pyrex™ Borosilicate Glass Narrow Neck Round Bottom Boiling Flask
Product Code. 10079139
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
4 L
6 L
10 L
20 L
Diameter (Metric) Outer:
50 mm
65 mm
85 mm
105 mm
131 mm
166 mm
207 mm
236 mm
279 mm
345 mm
Unit Size:
Each
Pack of 10
Product Code. 10079139
Brand: Pyrex 1100/32D
Only null left
Add to basket
Add to basket
Resistant to chemical corrosion and thermal shock
  • Complies with ISO 1773
  • Manufactured from Pyrex™ borosilicate glass which offers excellent resistance to chemical corrosion and thermal shock
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 1773
Capacity (Metric) 4 L
Material Borosilicate Glass
Height (Metric) 315 mm
Bottom Shape Round
Diameter (Metric) Outer 207 mm