Learn More
PX20606 trans racemate, MedChemExpress
Product Code. 16480649
Encompass_Preferred
Click to view available options
Quantity:
1 mg
2 mg
Unit Size:
1mg
2mg
Product Code. 16480649
Brand: Medchem Express HY100443A
MedChemExpress PX20606 trans racemate (PX-102 trans racemate) is a FXR agonist with EC50s of 32 and 34 nM for FXR in FRET and M1H assay, respectively.
PX20606 trans racemate (PX-102 trans racemate) is a FXR agonist with EC50s of 32 and 34 nM for FXR in FRET and M1H assay, respectively.
Specifications
Chemical Name or Material PX20606 trans racemate
CAS 1268244-85-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H22Cl3NO4
Quantity 1 mg
Synonym PX-102 trans racemate
Solubility Information DMSO : 100 mg/mL (180.23 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C([C@H]2[C@H](C3=CC=C(OCC4=C(C5CC5)ON=C4C6=C(Cl)C=CC=C6Cl)C=C3Cl)C2)C=C1
Molecular Weight (g/mol) 554.85
Formula Weight 554.85
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism