missing translation for 'onlineSavingsMsg'
Learn More

Ansell PX130 Lightweight Polyurethane-Coated Nylon Glove Liners

Product Code. 16805340 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16805340

Brand: Ansell C382070

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Polyurethane coating on the palm provides abrasion resistance, with an elasticated knit wrist to ensure a snug fit.

  • Polyurethane coating on the palm provides abrasion resistance
  • Elasticated knit wrist ensures a secure fit
  • Ventilated back is breathable to minimize perspiration
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Coated Area Palm
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A1
Hand Specific
Material Nylon
Size 7
For Use With (Application) Automotive Assembly, Machinery, and Equipment Operation
Glove Cuff Style Knit
Series PX130
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.