missing translation for 'onlineSavingsMsg'
Learn More

PWP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

483.00 EUR

Specifications

Antigen PWP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18248115
View Documents
Novus Biologicals
NBP1-79378
100 μL
483.00 EUR
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PWP1 Polyclonal specifically detects PWP1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

PWP1
Polyclonal
Rabbit
NP_008993
11137
Synthetic peptide directed towards the C terminal of human PWP1The immunogen for this antibody is PWP1. Peptide sequence FCSSCCPDLPFIYAFGGQKEGLRVWDISTVSSVNEAFGRRERLVLGSARN.
Primary
Western Blot
Unconjugated
RUO
IEF-SSP-9502, Keratinocyte protein IEF SSP 9502, nuclear phosphoprotein similar to S. cerevisiae PWP1, periodic tryptophan protein 1 homolog, PWP1 homolog (S. cerevisiae)
PWP1
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.