Learn More
Cole-Parmer™ Three-Way, Two-Position, Elliptic Ball Valves, NPT(F), PVDF
Description
- Easy disassembly for cleaning- simply pull out handle and push stem through bottom.
- Four-position valves are clearly marked with flow patterns to ensure correct handle position.
- O-rings made of Viton™, 316 SS handles, threaded connections, and nonrestrictive orifices.
- Rated up to 150 psi at 77°F and 230°F (110°C) at 50psi (liquids)
Specifications
Specifications
| Material | PVDF |
| For Use With | For 1/8 in. NPT connectors |
| For Use With (Application) | Liquid, gas, and vacuum applications |
| Packaging Quantity | 1 Pack |
| Product Line | Elliptic Ball Valve |
| Max. Operating Temperature (Metric) | 110°C |
| Product Type | Three-way Elliptic Ball Valve |