Learn More
Cole-Parmer™ PVDF Bulkhead Compression Union
Compression fittings are an excellent choice for connecting rigid tubing systems
Brand:  Cole-Parmer™ 06390-80
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11784809
  • 53.35 EUR / Each
Estimated Shipment: 05-12-2024
Description
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
  • Very good chemical resistance
  • Rated to 220psi maximum at 77°F ( 25°C)
  • Usable from 32 to 212°F (0 to 100°C)
Specifications
Specifications
Bulkhead Compression Union
Autoclavable
2 29/32 in.
1/2 in.
Autoclave or Ethylene Oxide
Compression Connection
PVDF
1/2 in.
1/2 in.
15/16 in.