Learn More
Cole-Parmer™ PVDF Bulkhead Compression Union
Compression fittings are an excellent choice for connecting rigid tubing systems
44.70 EUR - 53.35 EUR
Specifications
Product Type | Bulkhead Compression Union |
---|---|
Description | Compression Connection |
Autoclavable | Autoclavable |
Material | PVDF |
Sterilization Method | Autoclave or Ethylene Oxide |
Product Code | Brand | Port Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Port Size | Price | Quantity & Availability | |||||
11764809
|
Cole-Parmer™
06390-60 |
5/16 in. |
44.70 EUR
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
11784809
|
Cole-Parmer™
06390-80 |
1/2 in. |
53.35 EUR
Each |
Estimated Shipment: 22-11-2024
Log in to see stock. |
|||||
11774809
|
Cole-Parmer™
06390-70 |
3/8 in. |
52.60 EUR
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Very good chemical resistance
- Rated to 220psi maximum at 77°F ( 25°C)
- Usable from 32 to 212°F (0 to 100°C)
Specifications
Bulkhead Compression Union | |
Autoclavable | |
Autoclave or Ethylene Oxide |
Compression Connection | |
PVDF |