Learn More
Kautex™ PVC Transparent Rectangular Narrow Neck Bottles
Click to view available options
Capacity (Metric):
0.25 L
0.5 L
1 L
Unit Size:
Each
Pack of 106
Pack of 194
Pack of 390
Description
- Crystal clear transparent presentation
- Ripped sides for secure grip
- Leak-proof
- PVC
Order Info
Closures not included; sold separately.
Specifications
Specifications
| Capacity (Metric) | 1 L |
| Color | Transparent |
| Diameter (Metric) Inner Neck | 19.9 mm |
| For Use With (Application) | Storage, Packaging, Sending Samples, Mailing |
| Material | PVC |
| Neck Finish | Narrow |
| Closure Included | No |
| Mouth | Narrow |
| Shape | Square |
| Product Type | Narrow Neck Bottle |