Learn More
Cole-Parmer™ PVC Needle Valves
Specifications
Specifications
| Material | PVC |
| For Use With | 0.5 in. connection |
| Min. Operating Temperature (Metric) | 60°C |
| Packaging Quantity | 1 Pack |
| Max. Operating Temperature (Metric) | 60°C |
| Product Type | Needle Valve |