Learn More
Cole-Parmer™ PVC Needle Valves
Product Code. 11835342
Change view
Click to view available options
For Use With:
0.25 in. connection
0.5 in. connection
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
11835342 0.5 in. connection Each
11815342 0.25 in. connection Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11835342 Supplier Cole-Parmer™ Supplier No. 0324564
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Material PVC
For Use With 0.5 in. connection
Min. Operating Temperature (Metric) 60°C
Packaging Quantity 1 Pack
Max. Operating Temperature (Metric) 60°C
Product Type Needle Valve