Learn More
Cole-Parmer™ PVC Needle Valves
Product Code. 11815342
Change view
Click to view available options
For Use With:
0.25 in. connection
0.5 in. connection
Unit Size:
Each
Product Code. For Use With unitSize
11815342 0.25 in. connection Each
11835342 0.5 in. connection Each
2 options
Product Code. 11815342
Brand: Cole-Parmer™ 0324560
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Material PVC
For Use With 0.25 in. connection
Min. Operating Temperature (Metric) 60°C
Packaging Quantity 1 Pack
Max. Operating Temperature (Metric) 60°C
Product Type Needle Valve