missing translation for 'onlineSavingsMsg'
Learn More

Putrescine, ∽98%, MP Biomedicals™

Product Code. 11403130 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11403130

Brand: MP Biomedicals 0210044125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Putrescine is a GABA precursor in many biological systems and synthon for amido-ureas

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-60-1
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008235
InChI Key KIDHWZJUCRJVML-UHFFFAOYSA-N
Synonym 1,4-diaminobutane, putrescine, 1,4-butanediamine, tetramethylenediamine, butylenediamine, putrescin, 1,4-butylenediamine, tetramethyldiamine, 1,4-tetramethylenediamine, putrescina
PubChem CID 1045
ChEBI CHEBI:17148
IUPAC Name butane-1,4-diamine
SMILES C(CCN)CN

Specifications

Color Colorless
Density 0.877g/mL at 25°C
Assay Percent Range ∽98%
Quantity 25 g
Formula Weight 88.15
Physical Form Liquid
Chemical Name or Material Putrescine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.