missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Purple Screw Caps
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1000 Pk.
5000 Pk.
Unit Size:
Pack of 1000
Pack of 5000
Description
- Purple Caps fit empty FastPrep™ tubes and are used in FastPrep™ systems.
- Add sample to empty tube, add Lysing Matrix E material and lysing buffer, and apply cap. Process using FastPrep™ instrument or other bead-beater to lyse, grind or homogenize sample.
- Purple caps are also found in the Fast DNA™ Spin Kit for Soil and Fast RNA™ Pro Soil Kit.
Specifications
Specifications
| Color | Purple |
| Volume (Metric) | 2 mL |
| Storage Requirements | Store at Room Temperature (15°C to 30°C) |
| Quantity | 1000 Pk. |
| Product Type | Screw Cap |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction