Learn More
Fisherbrand™ Prefiltration Cartridge
Product Code. 15499639 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Cartridge, Activated Carbon
Cartridge, Anti-scaling Softening
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
15499639 Cartridge, Anti-scaling Softening Pack of 12
15489639 Cartridge, Activated Carbon Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15499639
Treats, softens, purifies, and conditions tap water.. Fisher Scientific™ Prefiltration Cartridges are used with the Accu500 Ultrapure Water System.
  • Anti-Scaling Softener Cartridge treats and conditions hard water
  • Activated Carbon Cartridge removes particles, organics and chlorine
Specifications
For Use With (Equipment) Accu500
Type Cartridge, Anti-scaling Softening
Height (Metric) 254 mm