Learn More
Labcon™ PurePlus™ Solid Frame PCR Plates
GREENER_CHOICE
Product Code. 30280324
Change view
Click to view available options
Array:
12 x 8
16 x 24
24 x 16
No. of Wells:
96
384
Volume (Metric) Well:
25 μL
0.1 mL
0.2 mL
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Array No. of Wells Volume (Metric) Well unitSize
30280324 24 x 16 384 25 μL Pack of 100
30280319 12 x 8 96 0.1 mL Pack of 100
30280314 12 x 8 96 0.2 mL Pack of 100
30280325 16 x 24 384 25 μL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30280324 Supplier Labcon™ Supplier No. 39875200009
Only null left
Add to basket
Add to basket
Labcon Solid Frame PCR plates are made with two materials to improve the fit in thermocyclers and allow for rapid cycling.
  • Made with a rigid polycarbonate frame enclosing polypropylene reaction wells
  • Available in styles to fit major cyclers and are freezable (-80°C)
  • Choose from 96 or 384 well capacity
Specifications
Barcode No Barcode
Certifications/Compliance ISO 9001:2015, FDA CFR 21 GMP
Format SBS Standard
Array 24 x 16
Autoclavable Not Autoclavable
Packaging 10 Plates/Pk., 10 Pk./Cs.
Color Clear
Disposable Single-use
Product Type PCR plate
Skirt Style Full skirt
Material Polycarbonate
Volume (Metric) Well 25 μL
No. of Wells 384
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) PCR
High-throughput Compatibility Yes
RNase Free Yes
Well Color Clear
Fit for Purpose PCR Ready