missing translation for 'onlineSavingsMsg'
Learn More

Veolia Water Systems Purelab™ Option S-7

Product Code. 13185960
Click to view available options
:
Flow Rate: <78L/hr
This item is not returnable. View return policy

Product Code. 13185960

Brand: Veolia Water Systems PRIPLB0220

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rich and robust Purelab™ Option-S Series systems use multiple purification technologies to produce general laboratory grade water at better than single distilled quality.

  • Purelab™ Option-S Series systems use multiple purification technologies to produce general laboratory grade water at better than single distilled quality
  • Purification technologies include pre-treatment, reverse osmosis and ion exchange
  • Applications include glassware washing/rinsing, feed to ultra pure water systems, reagent make-up/dilution, buffer and media preparation, general chemistry, qualitative analyses and feed to environmental cabinets

Specifications

Flow Rate <78 L/hr.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.