Learn More
Saint Gobain Life Sciences™ Pure Fit™ SIB™ Tees
Product Code. 17983431
Click to view available options
Diameter (Metric) Inner:
6.4 mm
9.5 mm
12.7 mm
Unit Size:
Pack of 25
Product Code. 17983431
Brand: Saint Gobain Life Sciences™ PFT500PPAF
Only null left
Add to basket
Add to basket
These tees utilize the patented BarbLock™ retention and seal system. Pure-Fit™ SIB™ provides a seamless transition that maintains fluid integrity and leak-free connections with complete biocompatibility for the most demanding applications and stringent test protocols.
  • Eliminates entrapment and leak points that can occur with traditional assembly systems
  • Unique internal design eliminates voids or dead space that can lead to turbulence or stagnation of fluid flow creating the potential for bacterial growth
  • Made of BPA-free, animal-free, gamma stable polypropylene
  • USP Class VI and ISO compliant
  • Manufactured and packaged in a certified clean room
  • Available in a variety of sizes
  • Total smooth inner bore (SIB) transition design
  • Complete 360° compression along the entire fitting system
  • No excessive voids at the interface or in the fluid path
  • Utilizes the patented BarbLock™ retention and seal system
  • Compatible with Saint-Gobain Sani-Tech™, C-Flex™ and PharMed™ flexible tubing
  • Applications include: Single-use Assemblies, Biopharmaceutical Manufacturing, Aseptic and Sterile Fill Lines, Filtration, General Laboratory, Final Fill, Product Sampling, Fluid Transfer
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Diameter (Metric) Inner 12.7 mm
Material Polypropylene
For Use With Biopharmaceutical Manufacturing, Single-use Assembly
Packaging Quantity 25 Tees/Pk.
Color Clear
Product Type Tubing Connector