Learn More
Watson-Marlow Tubing Pumpsil Platinum-Cured Silicone Tubing
Product Code. 30241002
Change view
Click to view available options
Diameter (Metric) Inner:
2.4 mm
6.4 mm
8.0 mm
Unit Size:
5m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
30241002 2.4 mm 5m
30241001 6.4 mm 5m
30241003 8.0 mm 5m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30241002 Supplier Watson-Marlow Tubing Supplier No. 913.AJ24.016
Only null left
Add to basket
Add to basket
Pumpsil is platinum-cured silicone tubing predominantly used for single-use biopharmaceutical applications, pH control and media feed in fermentation.
  • Developed for the biopharmaceutical industry, Pumpsil is exceptionally pure tubing offering an ultra-smooth bore which helps minimise protein binding and ensure high purity in the finished product.
  • Pumpsil is widely used for single-use biopharm applications, as well as pH control and media feed in fermentation, accurate metering, transfer and filtration. Absolute traceability with laser-etched part number, lot number and use-by-date. Fully documented biocompatibility and comprehensive validation pack.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Diameter (English) Inner 0.1 in.
Diameter (Metric) Inner 2.4 mm
Material Platinum-cured Silicone
Diameter (English) Outer 0.16 in.
Diameter (Metric) Outer 4 mm
Overall Length (English) 16 ft.
Overall Length (Metric) 5 m
Thickness (English) 0.0625 in.
Thickness (Metric) 1.6 mm
Min. Operating Temperature (English) -4°F
Min. Operating Temperature (Metric) -20°C
Color Clear
Hardness 60 (Shore A)
Flexibility Very Flexible
Max. Operating Temperature (English) 176°F
Max. Operating Temperature (Metric) 80°C
Product Type Tubing