Learn More
Masterflex PUMP,GEAR,MINI 12VDC
Brand:  Masterflex 07012-20
104.92 EUR valid until 2025-12-20
BEST PRICE on Promo! Use promo code "24121" to get your promotional price.
PUMP,GEAR,MINI 12VDC
Product Code. 12547416
  • 126.00 EUR / Each