Learn More
PTPRR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17937-1-AP-20UL
 View more versions of this product
Product Code. 16817954
  • 145.00 EUR / 20µL
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domains, and thus represents a receptor-type PTP. The similar gene predominately expressed in mouse brain was found to associate with, and thus regulate the activity and cellular localization of MAP kinases. The rat counterpart of this gene was reported to be regulated by the nerve growth factor, which suggested the function of this gene in neuronal growth and differentiation.
Specifications
Specifications
PTPRR
Polyclonal
Unconjugated
PTPRR
Ch 1PTPase, EC PTP, ECPTP, NC PTPCOM1, PCPTP1, PTP SL, PTPBR7, PTPRQ, PTPRR, R PTP R
Rabbit
Antigen Affinity Chromatography
RUO
19279, 5801, 94202
-20°C
Liquid
Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08617, Q15256, Q62132
PTPRR
PTPRR Fusion Protein Ag12145
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions