Learn More
Cytiva PTFE Unflanged Tubing
Shop All Cytiva Products
Click to view available options
Overall Length (Metric):
4 m
Unit Size:
Pack of 7
Specifications
Specifications
| Type | Unflanged Tubing |
| Diameter (Metric) Outer | 2.7 mm |
| Overall Length (Metric) | 4 m |
| Material | PTFE |