Learn More
Masterflex™ PTFE Solenoid Electric Valve
Product Code. 13553600
Change view
Click to view available options
Voltage:
12 VDC
24 VDC
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage unitSize
13553600 24 VDC Each
11710419 12 VDC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13553600 Supplier Masterflex™ Supplier No. 0135602
Only null left
Specifications
For Use With (Application) Suitable for pressure or vacuum service
Max. Operating Temperature (Metric) 70°C
Material PTFE
Quantity 1 Each
Type Solenoid Electric Valve
Voltage 24 VDC