Learn More
Buerkle™ Sealing Tape for PTFE Threaded Adapter
Product Code. 10790765 Shop All Bürkle Products
Click to view available options
For Use With:
Ball Valve
Unit Size:
Each
Product Code. 10790765
Brand: Buerkle™ 00064514
Only null left
Add to basket
Add to basket
Seals threads made of PTFE. Buerkle™ Sealing Tape for PTFE Threaded Adapter exhibits excellent chemical resistance, low abrasion, and is temperature resistant from -80°C to + 260°C.
Ideal for sealing threads made of PTFE. Features low abrasion, excellent chemical resistance, and temperature resistant from -80°C to + 260°C.
  • PTFE
  • Temperature resistant from -80°C to +260°C
TRUSTED_SUSTAINABILITY
Specifications
For Use With Ball Valve
Overall Height (Metric) 12 m
Overall Length (Metric) 1200 cm
Overall Width (Metric) 12 mm
Quantity 1 Roll
Type Sealing Tape