Learn More
Fisherbrand™ PTFE Micro Magnetic Stir Bar
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
3.50 EUR - 27.30 EUR
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
11898882
|
Fisherbrand™
|
1.5 mm | 8 mm |
9.25 EUR
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11868882
|
Fisherbrand™
|
3 mm | 3 mm |
10.85 EUR
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11808892
|
Fisherbrand™
|
3 mm | 13 mm |
7.67 EUR
Pack of 10 |
In Stock
Log in to see stock. |
||||
11878882
|
Fisherbrand™
|
2 mm | 5 mm |
15.05 EUR
Pack of 10 |
In Stock
Log in to see stock. |
||||
11828892
|
Fisherbrand™
|
1.5 mm | 15 mm |
14.10 EUR
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
12656316
|
Fisherbrand™
|
3 mm | 20 mm |
27.30 EUR
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
16245851
|
Fisherbrand™
|
3 mm | 30 mm |
17.00 EUR
Pack of 10 |
In Stock
Log in to see stock. |
||||
11888882
|
Fisherbrand™
|
3 mm | 6 mm |
6.16 EUR
Pack of 10 |
In Stock
Log in to see stock. |
||||
11858882
|
Fisherbrand™
|
2 mm | 2 mm |
12.40 EUR
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11818892
|
Fisherbrand™
|
3 mm | 8 mm |
12.07 EUR
Pack of 10 |
In Stock
Log in to see stock. |
||||
11888952
|
Fisherbrand™
|
2 mm | 7 mm |
6.16 EUR
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
10180301
|
Fisherbrand™
|
2 mm | 5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10652401
|
Fisherbrand™
|
3 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
- Supplied in reusable snap cap vials.
- For stirring in test tubes, small size beakers, or other containers.
- Autoclavable.