Learn More
RETSCH PTFE Grinding Jars
Product Code. 14831631 Shop All Retsch Products
Change view
Click to view available options
Capacity (Metric):
25 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
14831631 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 14831631 Supplier RETSCH Supplier No. 01.462.0335
Only null left
Specifications
Product Type Grinding Jar
Capacity (Metric) 25 mL
For Use With (Equipment) For CryoMill
Material PTFE