Learn More
Fisherbrand™ PTFE Centrifuge Test Tube with Closure
Product Code. 15913077 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
8 mL
13 mL
17 mL
45 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15913077 45 mL Each
10417172 13 mL Each
10581242 8 mL Each
10679122 17 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15913077
Only null left
Add to basket
Add to basket
Guaranteed inert and crack-free. Fisherbrand™ PTFE Centrifuge Test Tube with Closure can be used up to 280°C.
  • Isostatically molded in pure PTFE
  • Heavy wall construction
Specifications
Closure Color White
Color White
Capacity (Metric) 45 mL
Material PTFE
Quantity 1 Ea.
Autoclavable Autoclavable
Length (Metric) 150 mm
Closure Type Screw Cap
Diameter (Metric) 25 mm