Learn More
Cytiva PTFE Centering Ring
Description
- Description: Replacement Part for GV 050 Series Glass Vacuum Filtration Manifolds
- For Use With (Application): Vacuum Filtration
Specifications
Specifications
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| For Use With (Equipment) | Whatman GV050 Series Glass Vacuum Filtration System |
| Material | PTFE |
| Sterility | Non-sterile |
| For Use With (Application) | Vacuum Filtration |
| Model | GV050/0/05 |
| Quantity | 1 Ea. |
| Type | Centering Ring |