Learn More
Testo™ Pt100 Immersion/ Penetration Probe
Product Code. 12626067
Change view
Click to view available options
Length (Metric):
295 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
12626067 295 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12626067 Supplier Testo™ Supplier No. 0614 0235
Only null left
Add to basket
Add to basket
Ideal for applications in quality assurance, for calibration services and in laboratories
  • High-precision immersion/penetration probe (Pt100) with fixed cable including calibration protocol
  • Combined with a suitable Testo temperature measuring instrument, such as the testo 735, its system accuracy is so high that the measurement system can also be used as a working standard
  • Potential system accuracy of 0.05 °C at a resolution of 0.001 °C (depending on the measuring instrument connected)
  • Optional calibration to ISO or DAkkS possible
  • Measuring range: -80 to +300 °C
Specifications
Accuracy ±0.3°C (-80 to -40.001 °C), ±(0.1 °C + 0.05 % of mv) (-40 to 0.001 °C), ±0.05 °C (0 to +100 °C), ±(0.05 °C + 0.05 % of mv) (100.001 to 300 °C)
Response Time 60 sec.
Temperature Range (Metric) -80°C to +300°C
Length (Metric) 295 mm