Learn More
Testo™ Pt100 Immersion/ Penetration Probe
Description
- High-precision immersion/penetration probe (Pt100) with fixed cable including calibration protocol
- Combined with a suitable Testo temperature measuring instrument, such as the testo 735, its system accuracy is so high that the measurement system can also be used as a working standard
- Potential system accuracy of 0.05 °C at a resolution of 0.001 °C (depending on the measuring instrument connected)
- Optional calibration to ISO or DAkkS possible
- Measuring range: -80 to +300 °C
Specifications
Specifications
| Accuracy | ±0.3°C (-80 to -40.001 °C), ±(0.1 °C + 0.05 % of mv) (-40 to 0.001 °C), ±0.05 °C (0 to +100 °C), ±(0.05 °C + 0.05 % of mv) (100.001 to 300 °C) |
| Response Time | 60 sec. |
| Temperature Range (Metric) | -80°C to +300°C |
| Length (Metric) | 295 mm |