Learn More
RETSCH PT 200 Model Rotating Tube Dividers
Shop All Retsch Products
Click to view available options
Voltage:
100/240 V
220/240 V
Unit Size:
Each
Specifications
Specifications
| Product Type | Rotating Tube Divider |
| Dimensions (L x W x H) | 551 x 520 x 1050 mm |
| Electrical Requirements | 100/240 V, 50/60 Hz |
| For Use With (Application) | Agriculture, biology, chemistry/Plastics, construction materials, engineering/electronics, environment/recycling, food, geology/metallurgy, glass/ceramics, medicine/pharmaceuticals |
| Height (Metric) | 1050 mm |
| Hertz | 50/60 Hz |
| Length (Metric) | 551 mm |
| Model | PT 200 |
| Phase | 1-phase Power Connection |
| Voltage | 100/240 V |
| Show More |