missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA4 Monoclonal Antibody (OTI4B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504059
This item is not returnable.
View return policy
Description
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Three alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| PSMA4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P21670, P25789, Q9R1P0 | |
| Psma4 | |
| Full length human recombit protein of human PSMA4 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B8 | |
| Unconjugated | |
| PSMA4 | |
| HC9, HsT17706, PSC9 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 26441, 29671, 5685 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction