missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598411
This item is not returnable.
View return policy
Description
PSG3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The pregnancy-specific glycoproteins (PSGs) are a group of closely related secreted glycoproteins that are highly expressed in fetal placental syncytiotrophoblast cells. Members of the PSG protein family share a characteristic N-terminal domain that is homologous to the immunoglobulin variable region. PSGs become detectable in serum during the first two to three weeks of pregnancy and increase as the pregnancy progresses, eventually representing the most abundant fetal protein in the maternal blood at term. PSGs function to stimulate secretion of TH2-type cytokines from monocytes, and they may also modulate the maternal immune system during pregnancy, thereby protecting the semi-allotypic fetus from rejection. Eleven human PSG proteins (PSG1-PSG11) have been described.
Specifications
| PSG3 | |
| Polyclonal | |
| Unconjugated | |
| PSG3 | |
| Carcinoembryonic antigen SG5; pregnancy specific beta-1-glycoprotein 3; pregnancy-specific beta-1-glycoprotein 3; pregnancy-specific glycoprotein 3; PS-beta-G-3; PSBG-3; PSG3; sg5 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 5671 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q16557 | |
| PSG3 | |
| Recombinant Human Pregnancy-specific beta-1-glycoprotein 3 protein (35-310AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction