missing translation for 'onlineSavingsMsg'
Learn More

Pseudomonic Acid F Sodium Salt, TRC

Product Code. 30414261
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414261 25 mg 25mg
30411779 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30414261 Supplier Toronto Research Chemicals Supplier No. TRCP36029525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pseudomonic Acid F Sodium Salt
Analyte or Component Names Pseudomonic Acid F Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C24 H39 Na O9
Synonym Sodium 7-(((E)-4-((2S,3R,4R,5S)-3,4-Dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)heptanoate
InChI Formula InChI=1S/C24H40O9.Na/c1-14(11-21(28)31-9-7-5-4-6-8-20(26)27)10-18-23(30)22(29)17(13-32-18)12-19-24(33-19)15(2)16(3)25;/h11,15-19,22-25,29-30H,4-10,12-13H2,1-3H3,(H,26,27);/q;+1/p-1/b14-11+;/t15-,16-,17-,18-,19-,22+,23-,24-;/m0./s1
Solution Type Neat
SMILES C[C@H](O)[C@H](C)[C@@H]1O[C@H]1C[C@H]2CO[C@@H](C\C(=C\C(=O)OCCCCCCC(=O)O[Na])\C)[C@H](O)[C@@H]2O
IUPAC Name 7-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxyheptanoyloxysodium
Industry Type Pharmaceutical
Molecular Weight (g/mol) 494.551
Quantity 25 mg
Formula Weight 494.249
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.