Learn More
PRT062607 Hydrochloride, MedChemExpress
Product Code. 16472557
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16472557
Brand: Medchem Express HY15323
MedChemExpress PRT062607 Hydrochloride (P505-15 Hydrochloride) is a highly specific and potent inhibitor of purified Syk (IC50 1-2 nM).
PRT062607 Hydrochloride (P505-15 Hydrochloride) is a highly specific and potent inhibitor of purified Syk (IC50 1-2 nM).
Specifications
Chemical Name or Material PRT062607 Hydrochloride
CAS 1370261-97-4
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C19H24ClN9O
Quantity 50 mg
Synonym P505-15 Hydrochloride
Solubility Information H2O : ≥ 50 mg/mL (116.30 mM) ∣DMSO : ≥ 33 mg/mL (76.76 mM)
SMILES NC(C1=CN=C(N[C@H]2[C@@H](N)CCCC2)N=C1NC3=CC=CC(N4N=CC=N4)=C3)=O.[H]Cl
Molecular Weight (g/mol) 429.91
Formula Weight 429.91
Percent Purity 97.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease