missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRP19 Recombinant Rabbit Monoclonal Antibody (4M3Z1)
GREENER_CHOICE

Product Code. 17217073 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17217073 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17217073 Supplier Invitrogen™ Supplier No. MA538195

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, SKOV3, Mouse thymus, Rat spleen Immunogen sequence: FSENGYYLAT AADDSSVKLW DLRKLKNFKT LQLDNNFEVK SLIFDQSGTY LALGGTDVQI YICKQWTEIL HFTEHSGLTT GVAFGHHAKF IASTGMDRSL KFYSL.

PSO4 is the human homolog of yeast Pso4, a gene essential for cell survival and DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRP19
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 4M3Z1
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Prpf19
Gene Accession No. Q99KP6, Q9JMJ4, Q9UMS4
Gene Alias AA617263; AL024362; D19Wsu55e; hPSO4; Neuronal differentiation-related gene protein; NMP200; Nuclear matrix protein 200; nuclear matrix protein NMP200 related to splicing factor PRP19; nuclear matrix protein SNEV; pre-mRNA processing factor 19; pre-mRNA-processing factor 19; PRP19; prp19 beta; PRP19/PSO4 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae); PRPF19; PSO4; psoralen 4; RING-type E3 ubiquitin transferase PRP19; Senescence evasion factor; SNEV; UBOX4
Gene Symbols Prpf19
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 400-504 of human PRPF19(Q9UMS4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246216, 27339, 28000
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.