Learn More
Cytiva Protein and Peptide Molecular Weight Markers
Shop All Cytiva Products
Click to view available options
Product Type:
HMW Native Marker Kit
HMW-SDS Marker Kit
LMW-SDS Marker Kit
Unit Size:
Each
Description
- Get clear, sharp bands for easy measurement of migration distances
- Bands can be visualized by using Coomassie™ Brilliant Blue or silver staining
Specifications
Specifications
| Description | HMW-SDS Marker Kit |
| Color | Blue |
| Product Type | HMW-SDS Marker Kit |
| Form | Solid |
| For Use With (Application) | Designed for use with SDS electrophoresis |
| Kit Contents | Myosin: 25μg, 220000 (Mr), rabbit muscle (source), a-2-Macroglobulin: 100μg, 170000 (Mr), bovine plasma (source), b-Galactosidase: 16μg, 116000 (Mr), E. coli (source), Transferrin |
| Molecular Weight (g/mol) | Mr 220 000 |
| Quantity | 10 Vials |
| Storage Requirements | Unopened vials should be stored at 4°C |