Learn More
WTW™ Protective Armoring for WTW ProfiLine and MultiLine Portable Meters
Description
- Protective Armoring for ProfiLine and MultiLine Portable Meters
- For use with ProfiLine and MultiLine Portable Meters pH 3110
- ProfiLine/MultiLine Rubber arming for the Portable Meter ProfiLine pH 3110 that allows for easy and robust portable pH/mV meter for routine measurement
- Secure and reliable pH measurement due to repeatable results
- Accessory for the Portable Meter ProfiLine pH 3110
Specifications
Specifications
| For Use With (Equipment) | ProfiLine™ and MultiLine™ Portable Meters |
| Type | Protective Armor |
| Material | Rubber |