Learn More
SCHOTT AG Lighting and Imaging™ Protection Glass for LED Ring Light
Product Code. 12816553 Shop All Schott Products
Click to view available options
For Use With (Equipment):
EasyLED Ringlights or VisiLED Ringlights S80-25 or S80-55
Unit Size:
Each
Product Code. 12816553
Brand: SCHOTT AG Lighting and Imaging™ 400560
Only null left
Add to basket
Add to basket
Protection Glass for use with VisLED Brightfield Ring Lights, EasyLED Ring Light and EasyLED Ring Light Plus.
  • Borofloat™ glass protection window with adapting thread M110 x1.
  • Protects LEDs and microscope objective in oily and dusty environments.
  • Fits 66 mm ID.
Specifications
Type Protection Glass
Diameter (Metric) Inner 66 mm
For Use With (Application) Stereo Microscopy Illumination
For Use With (Equipment) EasyLED Ringlights or VisiLED Ringlights S80-25 or S80-55
Material Borofloat™ Glass
Size M110 x 1