missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protamine 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 EUR - 470.00 EUR
Specifications
| Antigen | Protamine 2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18331744
|
Novus Biologicals
NBP3-03223-20UL |
20 μL |
190.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18323225
|
Novus Biologicals
NBP3-03223-100UL |
100 μL |
470.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protamine 2 Polyclonal specifically detects Protamine 2 in Human samples. It is validated for Western Blot.Specifications
| Protamine 2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 5620 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| cancer/testis antigen family 94, member 2, CT94.2, FLJ27447, protamine 2, protamine-2, Sperm histone P2, Sperm protamine P2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-102 of human Protamine 2 (NP_002753.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title