missing translation for 'onlineSavingsMsg'
Learn More

PROTAC BET degrader-2, MedChemExpress

Product Code. 16474798
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474798 10 mg 10mg
16483246 25 mg 25mg
16437547 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474798 Supplier Medchem Express Supplier No. HY114228

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PROTAC BET degrader-2 is a PROTAC connected by ligands for Cereblon and BET with an IC50 value of 9.6 nM in cell growth inhibition in the RS4;11 cells and capable of achieving tumor regression.

PROTAC BET degrader-2 is a PROTAC connected by ligands for Cereblon and BET with an IC50 value of 9.6 nM in cell growth inhibition in the RS4

Specifications

Chemical Name or Material PROTAC BET degrader-2
CAS 2093388-33-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C41H42N10O6
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (64.86 mM; Need ultrasonic)
SMILES O=C(C1=NC(NC2=CC(C3CC3)=NN2CC)=C4C(NC5=C4C=C(OC)C(C6=C(C)ON=C6C)=C5)=N1)NCCCC7=CC=CC8=C7CN(C(CC9)C(NC9=O)=O)C8=O
Molecular Weight (g/mol) 770.84
Formula Weight 770.84
Percent Purity 98.19%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.