missing translation for 'onlineSavingsMsg'
Learn More

Propiconazole, TRC

Product Code. 30417610
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
Unit Size:
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417610 500 mg -
30409098 25 g 25g
30417486 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30417610 Supplier Toronto Research Chemicals Supplier No. TRCP770100500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60207-90-1
Molecular Formula C15 H17 Cl2 N3 O2
Molecular Weight (g/mol) 342.22
Synonym Propiconazole, 1H-1,2,4-Triazole, 1-[[2-(2,4-dichlorophenyl)-4-propyl-1,3-dioxolan-2-yl]methyl]-, 1-[[2-(2,4-Dichlorophenyl)-4-propyl-1,3-dioxolan-2-yl]methyl]-1H-1,2,4-triazole, 1-[2-(2,4-Dichlorophenyl)-4-n-propyl-1,3-dioxolan-2-ylmethyl]-1H-1,2,4-triazole, Aidol SW 900, Bamper, Bamper 25EC, Banner Maxx, Banner Maxx II, Binghuanzuo, Biogard PPZ, Biogard PPZ 250, Bumper, Bumper 250EC, Bumper 41.8EC, Busan 1292, CGA 64250, CGD 92710F, Cane Sett Treatment, Desmel, Dhan, Fertilome Liquid Systemic Fungicide, Juno, Juno (pesticide), Microban PZ, Microban S 2140, Mycostat P, Orbit, Orbit (fungicide), PPZ, PPZ (fungicide), Preventol A 12, Procon-Z, Procon-Z 14.3 L, Proconazole, PropiMax 3.6EC, Propiconazole, Propimax, Radar, Radar (insecticide), S 2140, Tilt, Tilt 125, Tilt 3.6EC, Tilt Premium, Wocosen, Wocosen Technical, Wocosin, Wocosin 50TK, Wolsit KD 10, Woodtreat 10
IUPAC Name 1-[[2-(2,4-dichlorophenyl)-4-propyl-1,3-dioxolan-2-yl]methyl]-1,2,4-triazole
SMILES CCCC1COC(Cn2cncn2)(O1)c3ccc(Cl)cc3Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H17Cl2N3O2/c1-2-3-12-7-21-15(22-12,8-20-10-18-9-19-20)13-5-4-11(16)6-14(13)17/h4-6,9-10,12H,2-3,7-8H2,1H3
Formula Weight 341.0698
Chemical Name or Material Propiconazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.