Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Propargite |
| CAS | 2312-35-8 |
| CAS Max % | ≤ 100% |
| Density | 1.12 g/cm3 (20°C) |
| Flash Point | 71.4°C |
| Beilstein | 6934289 |
| Color | Light Brown |
| Boiling Point | 210°C (Decomposition) |
| Synonym | 2-(4-tert-Butylphenoxy)cyclohexyl 2-propynyl sulfite |
| Molecular Weight (g/mol) | 350.47 g/mol |
| Show More |