missing translation for 'onlineSavingsMsg'
Learn More

Propane-1,3-diylbis((3-acetamidopropyl)carbamic Acid) di-tert-Butyl Ester, TRC

Product Code. 30417726
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417726 5 mg 5mg
30421913 2.5 mg 2.5mg
30417673 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30417726 Supplier Toronto Research Chemicals Supplier No. TRCP8352855MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propane-1,3-diylbis((3-acetamidopropyl)carbamic Acid) di-tert-Butyl Ester
Analyte or Component Names Propane-1,3-diylbis((3-acetamidopropyl)carbamic Acid) di-tert-Butyl Ester
Shipping Condition Room Temperature
Molecular Formula C23 H44 N4 O6
InChI Formula InChI=1S/C23H44N4O6/c1-18(28)24-12-9-14-26(20(30)32-22(3,4)5)16-11-17-27(15-10-13-25-19(2)29)21(31)33-23(6,7)8/h9-17H2,1-8H3,(H,24,28)(H,25,29)
Solution Type Neat
SMILES CC(=O)NCCCN(CCCN(CCCNC(=O)C)C(=O)OC(C)(C)C)C(=O)OC(C)(C)C
Industry Type Pharmaceutical
Molecular Weight (g/mol) 472.619
Quantity 5 mg
Formula Weight 472.326
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.