missing translation for 'onlineSavingsMsg'
Learn More

Pronethalol, MedChemExpress

Product Code. 16458688
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458688 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458688 Supplier Medchem Express Supplier No. HYB1238

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pronethalol ((±)-Pronethalo) is a non-selective β-adrenergic antagonist. Pronethalol is a potent inhibitor of Sox2 expression. Pronethalol protects against and to reverse Digitalis-induced ventricular arrhythmias and limits the cerebral arteriovenous malformation (AVMs).

Pronethalol ((±)-Pronethalo) is a non-selective β-adrenergic antagonist. Pronethalol is a potent inhibitor of Sox2 expression. Pronethalol protects against and to reverse Digitalis-induced ventricular arrhythmias and limits the cerebral arteriovenous malformation (AVMs).

Specifications

Chemical Name or Material Pronethalol
CAS 54-80-8
Color White
Purity Grade Notes Research
Molecular Formula C15H19NO
Quantity 10 mg
Synonym (±)-Pronethalo
Solubility Information DMSO : 50 mg/mL (218.04 mM; Need ultrasonic)
SMILES OC(CNC(C)C)C1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 229.32
Formula Weight 229.32
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.