missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolactin Antibody (rPRL/4909), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08245AF700
This item is not returnable.
View return policy
Description
Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Prolactin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| prolactin | |
| Recombinant fragment of human Prolactin (PRL) protein (around aa 63-201) (exact sequence is proprietary) (Uniprot: IDP01236) | |
| 100 μg | |
| Biologically Active Proteins, Cancer, Signal Transduction | |
| 5617 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rPRL/4909 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction