Learn More
WTW™ ProfiLine™ Oxi 3210 Portable Dissolved Oxygen Meter
Click to view available options
Calibration Modes:
1
Unit Size:
Each
Tekniske data
Tekniske data
| Includes | Meter with carry case including galvanic DO probe CellOx™ 325, cleaning solution, electrolyte, 3 exchange membrane heads, stand, beaker, instruction manual, CD-ROM and batteries |
| For Use With (Application) | For all portable field measurement applications |
| Pressure Range | 500 to 1100mbar |
| Calibration Modes | 1 |
| Temperature (Metric) Range | -5°C to +105°C |
| Resolution (Temperature) | 0.1 |
| Accuracy (Temperature) | ±0.1°C |
| Display Type | LCD |
| Data Points | 200 Manual points |
| Certifications/Compliance | cETLus, CE, ISO 9001:2008 |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion