Learn More
Productainer™ 3D BioProcess Container, BioTitan retainers, three ports, MPXs
GREENER_CHOICE
Product Code. 17938281 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
100 L
500 L
2000 L
Unit Size:
Pack of 5
Product Code. 17938281
Brand: Thermo Scientific SH31305.03
Specifications
Green Features 200 L
Capacity (Metric) 500 L
Film Description 100 L
For Use With (Equipment) 50 L
Quantity 200 L
Product Line 50 L
Tubing Style C-Flex