Learn More
IKA™ HABITAT Ferment, Double-walled 5 L Package
The HABITAT ferment package for the bioreactor HABITAT research contains a 5 L double-walled glass vessel for fermentation applications as well as the appropriate stirring drive.
Brand:  IKA™ 0010007661
Additional Details : Weight : 16.66000kg
Product Code. 17735089
  • POA / Each
Estimated Shipment: 20-05-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
Together with the separately available control tower package (e.g. HABITAT ferment) you receive all necessary components for a successful cultivation. For temperature control via the double jacket we recommend one of our our circulators such as the HRC 2 basic or control.
Scope of delivery
  • Stand 5
  • HA.gv.dw.5 Glass vessel, double-wall
  • HA.mt.s.10 Motor, big
  • HA.hp.s.5 Harvest pipe, straight
  • HA.ino Port for inoculation
  • HA.ba.5 Baffle
  • HA.sp.r.5 Ring sparger, 0.5 mm
  • HA.cn Condenser
  • HA.ip.b.5 6-blade segment impeller (2 pcs.)
  • HA.s.tm.5 Temperature sensor
  • HA.s.ph.5 pH sensor
  • HA.s.do.5 DO sensor
  • HA.s.fo Foam sensor
  • HA.s.lv.5 Level sensor
  • HA.cab.dw Cable and tube setHA.sf.250 Sample flask (4 pcs.)
Specifications
Specifications
Bioreactor Package
5 L
16.66 kg
Autoclavable
Borosilicate Glass