Learn More
Carbolite™ Furnace with Type R Temperature Sensor
Brand:  Carbolite™ RHF1603-230SN+&02-301
Additional Details : Weight : 100.00000kg
 View more versions of this product
Product Code. 10756904
  • 14336.50 EUR / Each
Estimated Shipment: 12-07-2024
Specifications
Compliance to BSEN 61010
3 L
Furnace
200 x 120 x 120 mm
120 mm
120 mm
200 mm
230 V
50/60 Hz
301 PID Controller
1600°C
For use in quality control, heat treatment and research of ceramic materials.
435 mm
655 mm
610 mm
230 V 50/60 Hz 4500 w
4500 w