missing translation for 'onlineSavingsMsg'
Learn More

Mouse Detection Antibody Diluent

Product Code. 15502117
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15502117 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15502117 Supplier Invitrogen Supplier No. EPX22222000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Dilution Buffer

TRUSTED_SUSTAINABILITY

Specifications

Product Type Diluent
Quantity 1 each
Species Mouse
For Use With (Equipment) Luminex™ Instruments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.