Learn More
Invitrogen™ eBioscience™ ProcartaPlex Mouse 36plex Control Set
Product Code. 15522497
Change view
Click to view available options
Quantity:
1 Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15522497 1 Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15522497 Supplier Invitrogen™ Supplier No. EPX36026092CTR
Contains a high and low control for all analytes. eBioscience™ ProcartaPlex Mouse 36plex Control Set is used with the ProcartaPlex Mouse Cytokine & Chemokine Panel 1A (36 plex).
eBioscience™ ProcartaPlex Mouse Cytokine & Chemokine Panel 1A (35 plex) (Mfr. Part No. EPX360-26092-901) is the ideal solution for assessing multiple protein biomarkers in a single sample. The assays can quantitate multiple targets in cell/tissue culture supernatants, serum and plasma using Luminex™ xMAP technology for the detection of secreted proteins including cytokines, chemokines, growth factors and other proteins.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Multiplex Control Set
Quantity 1 Pk.
Species Mouse
For Use With (Application) Multiplex Immunoassay
For Use With (Equipment) Luminex™ Instruments
Product Line ProcartaPlex
Protein Subtype CCL2 (MCP-1), CCL5 (RANTES), CCL7 (MCP-3), CXCL1 (GRO alpha), CXCL10 (IP-10), CXCL2 (MIP-2), ENA-78, Eotaxin, G-CSF, GM-CSF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-10, IL-12, IL-13, IL-15/IL-15R complex, IL-17A, IL-18, IL-2, IL-22, IL-23, IL-27, IL-28, IL-3, IL-31, IL-4, IL-5, IL-6, IL-9, LIF, M-CSF, MIP-1 alpha, MIP-1 beta, TNF alpha